missing translation for 'onlineSavingsMsg'
Learn More

Nickel ammonium sulfate hexahydrate, 98%

Catalog No. AAA1844122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1844122 100 g
AAA1844136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1844122 Supplier Thermo Scientific Chemicals Supplier No. A1844122

CAS: 7785-20-8 | H8N2NiO8S2·6H2O

Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 7785-20-8
Molecular Formula H8N2NiO8S2·6H2O
MDL Number MFCD00150260
Synonym Ammonium nickel sulfate hexahydrate

Specifications

Quantity 100 g
Linear Formula Ni(NH4)2(SO4)2·6H2O
UN Number UN3077
Merck Index 14,6501
Solubility Information Soluble in water.
Formula Weight 394.99 (286.90 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.923
Chemical Name or Material Nickel ammonium sulfate hexahydrate
Hazard Category H302+H332-H317-H334-H341-H350i-H360D-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 239-793-5
RTECSNumber WS6061000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only