Learn More
Innovatek Nichrome Innoculating Needle
Innovatek Nichrome Innoculating Needle with 8 in. Insulated Handle
Supplier: Innovatek 40303060
Description
- Nichrome wire offers all the benefits of platinum at much more reasonable cost
- Calibration gauge checks that inner diameter remains accurate during daily use
- Accurately calibrated with Evans Blue dye test
- Calibration gauge for quality control in the lab
- Consistent and reproducible results
- Rapid cooling
Specifications
| Inoculating Needle | |
| 24 G | |
| 8 in. | |
| 1 Needle/Pk. |