Learn More
Innovatek Nichrome Innoculating Loops
Catalog No. 361045037
Change view
Click to view available options
Gauge:
24 G
26 G
Calibrated:
No
Yes
Length (English) Handle:
No Handle Included
6 in.
8 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Calibrated Length (English) Handle
361045037 24 G Yes 8 in.
361045035 24 G No No Handle Included
361045036 26 G Yes 6 in.
361041862 26 G Yes No Handle Included
361045034 26 G No 8 in.
361045561 26 G No No Handle Included
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 361045037 Supplier Innovatek Supplier No. 40303740
Innovatek Nichrome Innoculating Loops glide smoothly across the agar surface for evenly isolated colonies
  • A wide range of premium quality, accurately sized twisted Nichrome wire loops
  • Nichrome wire offers all the benefits of platinum at much more reasonable cost
  • Accurately formed loops glide smoothly across the agar surface for evenly isolated colonies
  • Exacting manufacturing process ensures a perfectly symmetrical loop with equal dimensions, rather than an uneven oval
  • Calibration gauge checks that inner diameter remains accurate during daily use
  • Consistent and reproducible results
  • Accurately calibrated with Evans Blue dye test
  • Calibration gauge for quality control in the lab
  • Rapid cooling
Specifications
Disposable Reusable
Material Nichrome Wire
Gauge 24 G
Length (English) Handle 8 in.
Calibrated Yes