missing translation for 'onlineSavingsMsg'
Learn More

Niacinamide, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157896
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604792 500 g Poly Bottle
18604794 5 kg Poly Pail
18604795 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604792 Supplier Spectrum Chemical Mfg Cor Supplier No. NI105500GM
Only null left

NI105, 98-92-0, C6H6N2O

Spectrum™ Chemical Niacinamide, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 128-131°C
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98.5 to 101.5%
Grade USP