missing translation for 'onlineSavingsMsg'
Learn More

Niacinamide, FCC, 98.5-101%, Spectrum™ Chemical

Catalog No. p-7157895
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604787 100 g Poly Bottle
18604788 500 g Poly Bottle
18604789 1 kg Poly Pail
18604790 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604787 Supplier Spectrum Chemical Mfg Cor Supplier No. N1082100GM
Only null left

N1082, 98-92-0, C6H6N2O

Spectrum™ Chemical Niacinamide, FCC is a vitamin supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
InChI Key DFPAKSUCGFBDDF-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxamide
SMILES NC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 128-131°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98.5 to 101%
Grade FCC