missing translation for 'onlineSavingsMsg'
Learn More

Niacin, Powder, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18604785
Change view
Click to view available options
Quantity:
500 g
12 kg
25 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604785 50 kg Poly Bottle
18604781 500 g Poly Bottle
18604783 12 kg Poly Bottle
18604784 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604785 Supplier Spectrum Chemical Mfg Cor Supplier No. NI10050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NI100, 59-67-6, C6H5NO2

Spectrum™ Chemical Niacin, Powder, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 50 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Niacin Powder