missing translation for 'onlineSavingsMsg'
Learn More

Niacin, Powder, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7158474
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604780 100 g Poly Bottle
18604782 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604780 Supplier Spectrum Chemical Mfg Cor Supplier No. NI100100GM
Only null left

NI100, 59-67-6, C6H5NO2

Spectrum™ Chemical Niacin, Powder, USP is a vitamin supplement. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP