missing translation for 'onlineSavingsMsg'
Learn More

[NH2Me2][(RuCl((R)-xylbinap))2(mu-Cl)3], TCI America™

Catalog No. p-7134087
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1019200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1019200MG Supplier TCI America Supplier No. N1019200MG
Only null left

Chemical Identifiers

CAS 944451-08-5
Molecular Formula C106H107Cl5NP4Ru2
Molecular Weight (g/mol) 1898.31
MDL Number MFCD09753035
InChI Key LJSDQAUPXVWQNF-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(R)-(+)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131674757
IUPAC Name [chloraniumyl(chloro)ruthenio]chloranium dimethylamine bis({2'-[bis(3,5-dimethylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(3,5-dimethylphenyl)phosphane) hydrochloride ruthenium chloride
SMILES Cl.[Cl-].[Ru].CNC.[ClH+][Ru-]([ClH+])[ClH+].CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Quantity 200 mg
Formula Weight 1895.29
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((R)-xylbinap))2(mu-Cl)3]