missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-omega-Nitro-L-arginine-methyl ester hydrochloride, 98%

CAS: 51298-62-5 | C7H16ClN5O4 | 269.69 g/mol

$302.93 - $302.93

Chemical Identifiers

CAS 51298-62-5
Molecular Formula C7H16ClN5O4
Molecular Weight (g/mol) 269.69
MDL Number MFCD00039052,MFCD00133613
InChI Key QBNXAGZYLSRPJK-JEDNCBNOSA-N
Synonym h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
PubChem CID 135193
IUPAC Name methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328750050
SDS View Documents
Thermo Scientific Chemicals
328750050
5 g Glass Bottle
Each for $302.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51298-62-5
269.69
QBNXAGZYLSRPJK-JEDNCBNOSA-N
135193
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O
C7H16ClN5O4
MFCD00039052,MFCD00133613
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride

Specifications

51298-62-5
100.0
Authentic
C7H16ClN5O4
MFCD00039052,MFCD00133613
h-arg no2-ome.hcl, lname hydrochloride, l-name hydrochloride, ng-nitro-l-arginine methyl ester hydrochloride, nomega-nitro-l-arginine methyl ester hydrochloride, s-methyl 2-amino-5-3-nitroguanidino pentanoate hydrochloride, l-name, hcl, ng-no2-l-arg-ome, nomega-no2-l-arg-ome
[H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O
+ 14.50 (20.00°C c=3,CH3OH)
269.69
269.7
Crystalline Solid
97.5
White
98%
Glass Bottle
+14.0° to +16.0° (24°C, 589nm) (c=3, CH3OH)
QBNXAGZYLSRPJK-JEDNCBNOSA-N
methyl (2S)-2-amino-5-[[amino(nitramido)methylidene]amino]pentanoate;hydrochloride
5 g
135193
≥97.5%
N-ω-Nitro-L-arginine-methyl ester hydrochloride
Videos
SDS
Documents

Documents

RUO – Research Use Only