Learn More
Brady™ NFSI Certified Heavy Duty Anti-Skid Tape Roll - Silicon Carbide Grit, Solid Color, Black
Catalog No. 19103648
Change view
Click to view available options
Color:
Black
Catalog No. Color
19103648 Black
1 options
Catalog No. 19103648 Supplier Brady™ Supplier No. 110017
Only null left
This heavy-duty anti-skid tape is certified by the National Floor Safety Institute (NFSI) for high traction and wet slip resistance to help prevent slips, trips and falls.
  • Aggressive adhesive withstands harsh elements such as oil, fuel and grease
  • Durable high slip resistance with deeply textured silicon carbide grit
  • Anti-skid tape helps prevent slips, trips and falls in wet or high traffic areas
  • Ideal for use on loading docks, ramps and more
Specifications
Color Black