Learn More
SignalChem Biotech IkBA Protein 100ug
Catalog No. 502574633
Encompass
Recombinant human IkBA (1-175) was expressed in E. coli cells using an N-terminal GST tag.
Catalog No. 502574633 Supplier SignalChem Biotech Supplier No. I2031G100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human IkBA (1-175) was expressed in E. coli cells using an N-terminal GST tag.