missing translation for 'onlineSavingsMsg'
Learn More

Lime Oil, Distilled, FCC, 0.5%-2.5%, Spectrum™ Chemical

Catalog No. 18603897
Change view
Click to view available options
Quantity:
15 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603897 15 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18603897 Supplier Spectrum Chemical Mfg Cor Supplier No. L108615KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1086, 8008-26-2

Spectrum™ Chemical Lime Oil, Distilled, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8008-26-2

Specifications

CAS Max % 100%
Boiling Point 182°C/359.6°F
Packaging Amber Glass Bottle
Refractive Index 1.474-1.477
Quantity 15 kg
UN Number UN1197
Percent Purity 0.5 to 2.5%
Grade FCC
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Lime Oil, Distilled