Learn More
DuPont™ ProShield™ 60 Coverall (all)
Coveralls with laydown collar, open wrists and open ankles provide protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.
$205.24 - $205.24
Specifications
Product Type General Purpose Coveralls
Gender Unisex
Material ProShield
Color White
Disposable Single-use
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-088-2143
View Documents
DuPont™
NG120SWH4X002500
4X-Large
Case of 25 Each for $205.24
Only null left
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Laydown collar
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
Specifications
Specifications
General Purpose Coveralls
ProShield
Single-use
Open
Unhooded
Serged
Particle, Liquid
ASTM D751, D1117, D5034, D257, AATCC 127
Yes
10 mil
Unisex
White
Open
Collared
Front Zipper
Non-sterile
Laydown
Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
ProShield™ 60
Automotive Refinishing, Waste Cleanup, Remediation
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'