missing translation for 'onlineSavingsMsg'
Learn More

Sartorius NexaGel STEM

Catalog No. 14558128
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14558128 2 mL
14558127 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14558128 Supplier Sartorius Supplier No. NGH02S
Only null left

Xeno-free hydrogel system designed to enhance the performance of three-dimensional (3D) static suspension cultures.

  • Facilitate the scale-up of human pluripotent stem cells (hPSCs), creating a high-throughput platform for modeling various tissues and disease states.
  • This ready-to-use hydrogel system features an optimized formulation that supports the rapid expansion of high-quality 3D stem cell spheroids with pluripotent characteristics.
  • hPSCs can be directly thawed from liquid nitrogen or transferred from 2D matrix-coated culture vessels and immediately mixed with the hydrogel solution for static suspension cultures.
  • The optimized protocol is particularly beneficial for time-sensitive experiments, as it minimizes the need for frequent medium exchanges, thereby saving time and resources.
  • This hydrogel system is compatible with most hPSC culture media and tissue culture vessels.
  • The unique static suspension culture method eliminates the need for microcarriers in large-scale bioreactors, simplifying and streamlining cell harvesting.
  • The 3D stem cell spheroids generated using this system can be utilized for further sub-culturing, patterned differentiation, organoid development, or re-establishing 2D culture morphologies.

Specifications

Cell Type hPSCs
Form Liquid
Product Type Hydrogel System
Quantity 2 mL