Learn More
Sartorius NexaGel Hydrogel Matrix
Catalog No. 14558126
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14558126 2 mL
14558125 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14558126 Supplier Sartorius Supplier No. NGH01S
Only null left
Pre-prepared, xeno-free functional hydrogel ideal for 3D cell culture research.
  • This hydrogel is formulated with a precise combination of multi-functional ligands and concentrations, making it suitable for a wide variety of cell types and applications.
  • It can be directly combined with cell suspension without the need for further adjustments, providing a straightforward and versatile solution.
  • NexaGel™ Hydrogel Matrix effectively mimics the natural extracellular matrix (ECM), creating a more familiar environment for cells.
  • The hydrogel is stable at room temperature, maintains a neutral pH, is transparent, permeable, and works well with different imaging systems.
  • It converts into a hydrogel matrix by simply mixing with the cell culture medium.
  • Cells grown in this system can be easily retrieved using our NexaGel™ Cell Recovery Solution.
Specifications
Form Liquid
Product Type Hydrogel Matrix
Quantity 2 mL