Learn More
United Scientific Supplies Newtonian Demonstrator
Catalog No. S05908
Change view
Click to view available options
Newtonian demonstrator
Catalog No.
S05908 Newtonian demonstrator
1 options
Catalog No. S05908 Supplier United Scientific Supplies Supplier No. NWDM01
Illustrates Newton's Third Law of Motion and the principle of Conservation of Momentum.
  • Five polished metal balls with thin bifilar suspensions on a unique support design
Order Info
Shipping weight: 1.15 lb. (0.5kg); dimensions: 5.5 x 6 x 7 in. (14 x 15.2 x 17.8cm).
Specifications
Product Type Newtonian Demonstrator
For Use With (Application) Demonstrate Newton's third law of motion and the principle of conservation of momentum
Weight (English) 0.99 lb.
Weight (Metric) 0.44 kg