missing translation for 'onlineSavingsMsg'
Learn More

Learning Energy Bingo! Game

Catalog No. S01498
Change view
Click to view available options
Class Size:
30 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S01498 30 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01498 Supplier Customer Channels Group (Ccg) Supplier No. 246018

missing translation for 'validMainframeMsg'

Includes

30 full-color Bingo cards, 300 markers and a comprehensive teacher's guide.

The perfect means to review energy, its sources and uses.

  • Safe, clean and affordable sources of energy are critical for the future of our planet
  • Play Bingo using 75 standards based review cards
  • Students will be introduced to the basic vocabulary and concepts related to energy
  • They will be challenged to think, draw conclusions about energy resources and the consequences/challenges of using them
  • One side features a color photograph and key information about a specific energy concept
  • The cards can also be used as standalone flash cards for independent/small group study
  • Printed in color on durable, heavy cardstock

Specifications

For Use With (Application) Learn the basic vocabulary and concepts related to energy
Includes 75 standards based review cards, 30 full-color Bingo cards, 300 markers and a comprehensive teacher's guide
Class Size 30 Students
Description Energy Bingo Game