missing translation for 'onlineSavingsMsg'
Learn More

NewPath Learning™ Life Science Curriculum Mastery Game

Catalog No. S95684
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95684 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95684 Supplier NewPath Learning™ Supplier No. 246007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combine Game-Based Learning with Online Resources for Engaging Review and Reinforcement

Use game-based learning to build student skills on core curriculum concepts. Includes enough materials for an entire class of up to 36 students including 750 illustrated, self-correcting review questions – 30 game cards for each of 25 topics; 12 double-sided, laminated game boards; game pieces and Teacher's guide. Also includes a 1 Year Premium Subscription to NewPath's Online Learning Program providing access to standards-based online Science resources including animated/narrated multimedia lessons; interactive exercises; virtual labs; quizzes; games and more.

Complete curriculum coverage of the following standards-based topics –

  • The World of Life Science
  • Diversity of Life
  • Cells: The Basic Units of Life
  • Cell Processes
  • Cell Reproduction
  • Genetics – Study of Heredity
  • Modern Genetics
  • The Evolution and Interaction of Living Things
  • Bacteria and Viruses
  • Protists and Fungi
  • Introduction to Plants
  • Plant Processes
  • Plant Reproduction
  • Introduction to Animals
  • Sponges, Cnidarians and Worms
  • Mollusks, Arthropods and Echinoderms
  • Fishes, Amphibians and Reptiles
  • Birds and Mammals
  • Bones, Muscles and Skin
  • The Digestive System and Nutrition
  • Circulation and Immunity
  • Respiration and Excretion
  • The Nervous System
  • The Endocrine System and Reproduction
  • Ecosystems, Food Chains and Food Webs

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 5 to 12
Format Game
Product Type Life Science game
Includes 750 Review cards, teachers guide, classroom presentation CD
For Use With (Application) Comprehensive coverage of the current national science education standards
Subject Science