missing translation for 'onlineSavingsMsg'
Learn More

New Pig Hazardous Materials Disposal Bags

Catalog No. 189995701
Change view
Click to view available options
Capacity (English):
45 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
189995701 45 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 189995701 Supplier New Pig Supplier No. BAG202L

missing translation for 'validMainframeMsg'

Confidently bag hazardous or contaminated waste for disposal.

  • Tough polyethylene bags resist punctures, tearing and blowouts
  • Durable 3 mil thickness
  • Bright yellow bags are boldly labeled Hazardous Material - Handle With Care

Specifications

Capacity (Metric) 170.3 L
Capacity (English) 45 gal.
Color Yellow, Black
Height (Metric) 152.4 cm
Width (Metric) 91.44 cm
For Use With (Application) Ideal for lining 30 and 55 gal. drums or for temporary storage of saturated absorbents and other waste
Width (English) 36 in.
Thickness 3 mil
Height (English) 60 in.
Material Polyethylene
Product Type Hazardous Disposal Bag
Show More Show Less