missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Methylene Blue N, zinc chloride double salt, pure

Catalog No. ac192020100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC192020100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC192020100 Supplier Thermo Scientific Chemicals Supplier No. 192020100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0·5 ZnCl2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula 0·5 ZnCl2
MDL Number MFCD00078135
Synonym Basic Blue 24, C.I. 52030

Specifications

Absorbance (c=5mg/l; water), (c=5mg/l; water) at 0nm
Infrared Spectrum Authentic
Lambda Max 628 to 634nm, Molar Absorption Coefficient 40000 min., 588 to 594nm, Molar Absorption Coefficient 36000 min., 284 to 290nm, Molar Absorption Coefficient 28000 min., 242 to 248nm, Molar Absorption Coefficient 16000 min.
Packaging Glass bottle
Color Brown to Purple
Quantity 10 g
Physical Form Crystalline Powder
Formula Weight 416.04
Grade Pure
Chemical Name or Material New Methylene Blue N, zinc chloride double salt

RUO – Research Use Only