missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Coccine, pure

Catalog No. AC190231000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190231000 100 g Glass bottle
AC190230250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190231000 Supplier Thermo Scientific Chemicals Supplier No. 190231000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2611-82-7 | C20H11N2Na3O10S3 | 604.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2611-82-7
Molecular Formula C20H11N2Na3O10S3
Molecular Weight (g/mol) 604.46
MDL Number MFCD00004084
InChI Key IVNZBWNBYXERPK-DZGBHZPSSA-K
Synonym Acid Red 18, C.I. 16255, Cochineal Red A
PubChem CID 131851859
IUPAC Name (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Specifications

Dye Content 75% min.
Infrared Spectrum Authentic
Loss on Drying 10% max. (135°C, 6 hrs)
Packaging Glass bottle
UV Spectrometry Complies
Beilstein 16, I, 306
Color Bordeaux to Brown
Quantity 100 g
Physical Form Powder
Formula Weight 604.46
Grade Extra Pure
Chemical Name or Material New Coccine
Show More Show Less
EINECSNumber 220-036-2
RTECSNumber QJ6530000
TSCA TSCA

RUO – Research Use Only