Learn More
Eppendorf™ Test Tube Racks for Model C76 Water Bath Shaker
Catalog No. p-137785
Change view
Click to view available options
Holds:
18.0 25 mm Tubes
29.0 16 mm Tubes
36.0 20 mm Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
1428424 36.0 20 mm Tubes
05414375 18.0 25 mm Tubes
05414374 29.0 16 mm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1428424 Supplier Eppendorf™ Supplier No. AG7TT20
Only null left
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Require subplatform (14-284-46); order separately.
Specifications
For Use With (Equipment) C76 water bath shaker
Holds 36.0 20 mm Tubes
Includes Insert
Material Stainless Steel