Learn More
Eppendorf™ Scientific Shaker Test Tube Platforms
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Supplier:  Eppendorf™ M12890300
 View more versions of this product
Catalog No. 14285698
Only null left
Description
Description
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Chemical Identifiers
Test Tube Rack
Stainless Steel
Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series
Large
Specifications
15 to 18 mm
Stainless Steel
42
Test Tube Rack
Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series
Large
Product Suggestions
Product Suggestions