Learn More
Eppendorf™ Scientific Shaker Test Tube Platforms
Description
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
Specifications
Specifications
| Tube Size | 12 to 15 mm |
| Product Type | Test Tube Rack |
| Material | Stainless Steel |
| For Use With (Equipment) | Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series |
| No. of Positions | 60 |
| Size | Large |