missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine 98.0+%, TCI America™

Catalog No. N0922200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0922200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0922200MG Supplier TCI America Supplier No. N0922200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nevirapine is a non-nucleoside reverse transcriptase inhibitor intended for research applications.

Chemical Identifiers

CAS 129618-40-2
Molecular Formula C15H14N4O
Molecular Weight (g/mol) 266.304
MDL Number MFCD00866928
InChI Key NQDJXKOVJZTUJA-UHFFFAOYSA-N
Synonym nevirapine, viramune, bi-rg-587, viramune xr, nvp, birg587, 11-cyclopropyl-4-methyl-5,11-dihydro-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one, nevirapine usan:inn, unii-99dk7fvk1h, 11-cyclopropyl-5,11-dihydro-4-methyl-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one
PubChem CID 4463
ChEBI CHEBI:63613
IUPAC Name 11-cyclopropyl-4-methyl-5H-dipyrido[2,3-e
SMILES CC1=C2C(=NC=C1)N(C3=C(C=CC=N3)C(=O)N2)C4CC4

Specifications

Melting Point 246°C
Color White-Yellow
Quantity 200 mg
Formula Weight 266.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nevirapine
RTECSNumber JM5562500
TSCA No
Recommended Storage Freezer