missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Neutral Red, pure, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 290.0°C |
| Absorption Ratio | 1 to 1.12 |
| Dye Content | 50% min. |
| Infrared Spectrum | Authentic |
| Lambda Max | 539 to 544nm |
| Packaging | Glass bottle |
| Beilstein | 25, 401 |
| Merck Index | 15, 6573 |
| Color | Green |
| Solubility Information | Solubility in water: 50g/L. Other solubilities: 1.8% in abs. alcohol, 3.75% in cellosolve, 3.0% in ethylene glycol, practically insoluble in xylene |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Suspected of causing genetic defects. |
| Precautionary Statement | GHS P Statement: Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge |
RUO – Research Use Only