missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol

Supplier:  Thermo Scientific Chemicals J6305406

Quantity 5 g

 View more versions of this product

Catalog No. AAJ6305406


Only null left

Description

Description

Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

553-24-2
288.779
PGSADBUBUOPOJS-UHFFFAOYSA-N
11105
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
C15H17ClN4
MFCD00012651
Basic Red 5; C.I. 50040
CHEBI:86370
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Neutral Red
553-24-2
MFCD00012651
14,6488
PGSADBUBUOPOJS-UHFFFAOYSA-N
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
11105
288.78
5 g
290°C (decomposition)
C15H17ClN4
3918740
Basic Red 5; C.I. 50040
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
288.779
CHEBI:86370
Black to Green
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-035-8

RTECSNumber : SG1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions