Learn More
StatLab™ Neutral Buffered Formalin 20%
Catalog No. STL286205
Change view
Click to view available options
Quantity:
5 gal.
Catalog No. Quantity
STL286205 5 gal.
1 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL286205 Supplier StatLab™ Supplier No. 286205*
Only null left
Harmful if swallowed, if in contact with skin, or if inhaled.
20% NBF intended as a general fixative for all tissues to be examined in light microscopy.
  • Often used for surgical and necropsy specimens
  • Ideal solution for brain tissue
Specifications
Packaging Type Cube
Quantity 5 gal.
For Use With (Application) Tissue Fixation