Learn More
Neutec PS200 Automated Agar Filler
Catalog No. 14920577
Change view
Click to view available options
PS200
Catalog No.
14920577 PS200
1 options
Catalog No. 14920577 Supplier Neutec Supplier No. NT BT950000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
For Use With (Equipment) 89-96 mm dish dia./ 14-20.5 mm H
Manual/Automated Automated
Transfer Volume 1.0-99.9 mL