Learn More
Hamilton™ Neuros™ 1700 Series Gas Tight Syringes with Removable Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Supplier:  Hamilton™ 6546005
 View more versions of this product
Catalog No. 14815907
Only null left
Description
Description
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Chemical Identifiers
Autoclavable
0.004 in.
0.008 in.
Controlled animal injections
Gastight
239°F
50°F
33 G
0.108 mm
0.210 mm
0 to 20 mm
1000 psig (68.9 bar)
115°C
Specifications
Autoclavable
33 G
0.004 in.
0.008 in.
0.460 mm
0.210 mm
Neuros™ adapter, two adjustable needle sleeves (one with a blind stop and one without)
1 in.
Gastight
239°F
50°F
3
Ea.
Removable Blunt
Borosilicate Glass, Borosilicate Glass (Barrel), PTFE (Plunger), PTFE (Tip)
0.018 in.
0.108 mm
0.260 in.
6.60 mm
Controlled animal injections
1 in.
0 to 20 mm
1000 psig (68.9 bar)
115°C
1701 RN
10°C
10 μL
Product Suggestions
Product Suggestions