Learn More
Hamilton™ Neuros™ 1700 Series Gas Tight Syringes with Removable Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Supplier:  Hamilton™ 6546016
 View more versions of this product
Catalog No. 14785712
Only null left
Description
Description
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Chemical Identifiers
33 G
0 to 20 mm
1000 psig (68.9 bar)
115°C
Controlled animal injections
Gastight
239°F
50°F
Specifications
Borosilicate Glass, PTFE (Plunger)
Controlled animal injections
Gastight
239°F
50°F
4
33 G
0 to 20 mm
1000 psig (68.9 bar)
115°C
1705 RN
Product Suggestions
Product Suggestions