missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neuropeptide Y, Porcine

Catalog No. AAJ66933MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66933MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66933MCR Supplier Thermo Scientific Chemicals Supplier No. J66933MCR

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Porcine
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 4253.70
Product Type Neuropeptide Y, porcine
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence NPY

RUO – Research Use Only