missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Neuropeptide FF

Catalog No. AAJ66509MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66509MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66509MCR Supplier Thermo Scientific™ Supplier No. J66509MCR

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Purification Method HPLC
Common Name F-8-F-NH2
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1081.29
Product Type Neuropeptide FF
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Phe-Leu-Phe-Gln-Pro-Gln-Arg-Phe-NH2

For Research Use Only