Learn More
Stemcell Technologies Inc NeuroCult™ NS-A Basal Medium (Human), Size: 450 mL
Catalog No. NC1290682
Encompass
NeuroCult™ NS-A Basal Medium (Human) is a standardized basal medium for the expansion of human neural stem cells and brain tumor stem cells, in the neurosphere or adherent monolayer culture system. This basal medium is a component of the NeuroCult™ NS-A Proliferation Kit (Human; Catalog #05751) and the NeuroCult™ NS-A Differentiation Kit (Human; Catalog #05752).
Catalog No. NC1290682 Supplier Stemcell Technologies Inc Supplier No. 05750
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NeuroCult™ NS-A Basal Medium (Human) is a standardized basal medium for the expansion of human neural stem cells and brain tumor stem cells, in the neurosphere or adherent monolayer culture system. This basal medium is a component of the NeuroCult™ NS-A Proliferation Kit (Human; Catalog #05751) and the NeuroCult™ NS-A Differentiation Kit (Human; Catalog #05752).