missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuraminidase, Clostridium perfringens, 50% w/w sucrose, Thermo Scientific Chemicals
$335.41 - $593.14
Chemical Identifiers
| CAS | 9001-67-6 |
|---|---|
| MDL Number | MFCD00131711 |
| Synonym | Acyl-neuraminyl hydrolase,EC 3.2.1.18 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60711CPL
|
Thermo Scientific Chemicals
J60711CPL |
5 U |
Each for $335.41
|
|
|||||
|
AAJ60711LR
|
Thermo Scientific Chemicals
J60711LR |
10 U |
Each for $593.14
|
|
|||||
Description
A hydrolytic enzyme that removes sialic acid from mucoproteins
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9001-67-6 | |
| MFCD00131711 | |
| Acyl-neuraminyl hydrolase,EC 3.2.1.18 |
Specifications
| 9001-67-6 | |
| MFCD00131711 | |
| Acyl-neuraminyl hydrolase,EC 3.2.1.18 | |
| Lyophilized Powder |
| Plastic bottle | |
| 5 U | |
| 50% w/w sucrose | |
| Neuraminidase |
Safety and Handling
EINECSNumber : 232-624-6
RTECSNumber : QQ3450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only