missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ NeuLog™ pH Logger Sensor
Description
- Sealed reference system and gel fill designed for ease of use and long life
- Fast response across the full pH range and can be calibrated with any standard buffer solution
- Duration: 1 second to 31 days
- Range: 0 to 14
- Resolution: 0.01pH
- Rate: 100 S/sec.
- Requires connection hardware, sold separately
Specifications
Specifications
| Type | Sensor, pH |
| Includes | Drop counter logger sensor, drop counter probe, guide |
| Certifications/Compliance | ISO 9001 |
| Grade | 3+ |
| For Use With (Application) | Measures the static pH of common liquids and changing values in titrations |
| For Use With (Equipment) | NeuLog neuron logger sensors |
| Range | 0 to 14 |
| Resolution | 0.01 |
| Dimensions (L x W x H) | 4 x 8.5 x 2 in. (10.16 x 21.6 x 5.08 cm) |
| Height (English) | 2 in. |
| Show More |