missing translation for 'onlineSavingsMsg'
Learn More

Neu5GcAc[1Me,478Ac,9N3]-β-SPh 93.0+%, TCI America™

Catalog No. m2696100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2696100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2696100MG Supplier TCI America Supplier No. M2696100MG
Only null left

Chemical Identifiers

CAS 1195053-25-8
Molecular Formula C26H32N4O12S
Molecular Weight (g/mol) 624.618
InChI Key YBDMZZOROXIIKI-YJGXBXLCSA-N
Synonym Methyl 5-Acetoxyacetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonate, Neu5GcAc[1Me,478Ac,9N3]-beta-SPh
PubChem CID 125128932
IUPAC Name methyl (2R,4S,5R,6R)-4-acetyloxy-5-[(2-acetyloxyacetyl)amino]-6-[(1R,2R)-1,2-diacetyloxy-3-azidopropyl]-2-phenylsulfanyloxane-2-carboxylate
SMILES CC(=O)OCC(=O)NC1C(CC(OC1C(C(CN=[N+]=[N-])OC(=O)C)OC(=O)C)(C(=O)OC)SC2=CC=CC=C2)OC(=O)C

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 624.62
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl (Phenyl 5-Acetoxyacetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-
TSCA No
Recommended Storage Refrigerator