missing translation for 'onlineSavingsMsg'
Learn More

Neu5Ac alpha(2-3)Gal beta(1-4)GlcNAc-beta-pNP 95.0+%, TCI America™

Catalog No. n08542mg
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08542MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08542MG Supplier TCI America Supplier No. N08542MG
Only null left

Chemical Identifiers

CAS 501427-92-5
Molecular Formula C31H45N3O21
Molecular Weight (g/mol) 795.701
InChI Key OUJZEVYPUZNYBL-SGZWRCNRSA-N
PubChem CID 44630015
IUPAC Name (2S,4R,5R,6R)-5-acetamido-2-[(2S,3S,4S,5S,6S)-2-[(2S,3S,4R,5S,6S)-5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3C(OC(C(C3O)NC(=O)C)OC4=CC=C(C=C4)[N+](=O)[O-])CO)CO)O)O

Specifications

Color White
Quantity 2 mg
Formula Weight 795.70
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac alpha(2-3)Gal beta(1-4)GlcNAc-beta-pNP
TSCA No
Recommended Storage Freezer