missing translation for 'onlineSavingsMsg'
Learn More

Neu5Ac[1Me,4789Ac]alpha(2-3)Gal[246Bz]-beta-MP 95.0+%, TCI America™

Catalog No. N0846200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0846200MG 200 mg
N08461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0846200MG Supplier TCI America Supplier No. N0846200MG

Chemical Identifiers

CAS 1858223-85-4
Molecular Formula C54H57NO22
Molecular Weight (g/mol) 1072.035
InChI Key DNICDVYFYXDKKN-KJIULPAQSA-N
Synonym 4-Methoxyphenyl 2,4,6-Tri-O-benzoyl-3-O-(methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonate)-beta-D-galactopyranoside
PubChem CID 121234060
IUPAC Name methyl (2S,4S,5R,6R)-5-acetamido-4-acetyloxy-2-[(2R,3S,4S,5R,6S)-3,5-dibenzoyloxy-2-(benzoyloxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-6-[(1S,2R)-1,2,3-triacetyloxypropyl]oxane-2-carboxylate
SMILES CC(=O)NC1C(CC(OC1C(C(COC(=O)C)OC(=O)C)OC(=O)C)(C(=O)OC)OC2C(C(OC(C2OC(=O)C3=CC=CC=C3)OC4=CC=C(C=C4)OC)COC(=O)C5=CC=CC=C5)OC(=O)C6=CC=CC=C6)OC(=O)C

Specifications

Color White
Quantity 200 mg
Formula Weight 1072.04
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac[1Me,4789Ac]alpha(2-3)Gal[246Bz]-beta-MP
TSCA No
Recommended Storage Refrigerator