Learn More
Ancare Corporation NESTLETS NESTING MATERIAL-MICE
Catalog No. NC9365966
Encompass
When ordering 1-4 cases, please order this part. Perfect nesting material for mice, hamsters, gerbils, rats, rabbits and other small animals. Chemically inert, odorless and non-ingestible. Made of pulped virgin cotton fiber clean, dust-free and autoclavable. Each carton (22 x 12 x 12) contains 100 sheets, 12 x 12, scored to break easily into a total of 3600 individual Nestlets, each 2 square.
Catalog No. NC9365966 Supplier Ancare Corporation Supplier No. NES3600QTY 14 CS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
When ordering 1-4 cases, please order this part. Perfect nesting material for mice, hamsters, gerbils, rats, rabbits and other small animals. Chemically inert, odorless and non-ingestible. Made of pulped virgin cotton fiber clean, dust-free and autoclavable. Each carton (22 x 12 x 12) contains 100 sheets, 12 x 12, scored to break easily into a total of 3600 individual Nestlets, each 2 square.