missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Nest and Tub RTU Format Velocity™ Vials

Catalog No. 01677093
Change view
Click to view available options
Product Line:
10R
2R
6R
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
01677093 2R
01677094 6R
01677095 10R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01677093 Supplier Corning™ Supplier No. 90388E5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Velocity™ vials in Nest and Tub RTU format.

Corning™ Velocity™ vials

  • Type I externally coated borosilicate glass vials
  • RTU with SG EZ-fill™ Technology
  • Nest and Tub RTU format

Specifications

Material Borosilicate Glass
Volume (Metric) Brim 4 mL
Product Line 2R
Height (Metric) 35 mm
Product Type Pharmaceutical Vial