missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Nest and Tub RTU Format Type I Glass Vials

Catalog No. p-200025530
Change view
Click to view available options
Product Line:
10R
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
01677078 10R
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01677078 Supplier Corning™ Supplier No. 80093E5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality Type I glass vials in Nest and Tub RTU format.

  • Type I uncoated borosilicate glass vials
  • RTU with SG EZ-fill™ Technology
  • Nest and Tub RTU format

Specifications

Material Borosilicate Glass
Volume (Metric) Brim 13.5 mL
Product Line 10R
Height (Metric) 45 mm
Product Type Pharmaceutical Vial