missing translation for 'onlineSavingsMsg'
Learn More

Nessler's Reagent (Mercuric - Potassium Iodide TS, Alkaline), USP 25 Formulation, Ricca Chemical

For USP 25 Testing

$534.17 - $728.97

Specifications

Color Clear light yellow
pH Range >12
Format Liquid
Packaging Amber Poly Bottle
Type Nessler's Reagent
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
525516
View Documents Promotion Details
N/A 500 mL
Each for $534.17
Only null left
 
525532
View Documents Promotion Details
N/A 1 L
Each for $728.97
Only null left
 
Description

Description

Specifications

Specifications

Clear light yellow
Liquid
Nessler's Reagent
>12
Amber Poly Bottle
Videos
SDS
Documents

Documents