missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NERL™ Reagent Grade Water

Catalog No. 23249581
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23249581 5 gal.
23249580 32 oz.
23249579 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23249581 Supplier Thermo Scientific™ Supplier No. J98005
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve control in critical diagnostic procedures by using this reagent grade water in place of deionized or distilled water.

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ Reagent Grade Water in place of deionized or distilled water. This water meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.

  • Ideal for reconstituting chemistry or coagulation products, preparing analytical standards, or rinsing delicate electrodes
  • Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron
  • Essentially free from organic and inorganic particulate and soluble contaminants
  • Use in place of deionized or distilled water to improve control in critical diagnostic procedures
  • Contains no preservatives
  • Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water
  • Certificate of Analysis is available online

Specifications

pH 5 to 7
Purity Grade Notes Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online.
Grade USP/NF, CLRW
Packaging Cubitainer
Quantity 5 gal.

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.