missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NERL™ Diluent 2 Hematology Reagent for Flow Cytometry

Catalog No. 23029361
Change view
Click to view available options
Quantity:
20 L Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23029361 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23029361 Supplier Thermo Scientific™ Supplier No. JDIL5522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hematology reagents for use with various Beckman Coulter& analyzers. Multi-Purpose Blood diluent.

 

Multi-Purpose Blood Diluent for use on Coulter Analyzers. Cut costs by replacing manufacturer's reagents with Thermo Scientific™ NERL™ Diluent 2 Hematology Reagent for Flow Cytometry. This anti-microbial additive keeps reagent purity constant, with no added detergents.

Specifications

Content And Storage Store in a cool, dry area at room temperature (15°-30°C).
Description Thermo Scientific NERL Diluent 2 Hematology Reagent
For Use With (Equipment) Coulter Analyzers not Performing WBC Differentials
Type Hematology Reagent
Quantity 20 L Cubitainer