missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NERL™ Blood Bank Saline

Catalog No. 23249605
Change view
Click to view available options
Quantity:
1 x 10 L
1 x 20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23249605 1 x 10 L
23332408 1 x 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23249605 Supplier Thermo Scientific™ Supplier No. J8505
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhance weak agglutination reactions with this saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood.

Enhance weak agglutination reactions with Thermo Scientific™ NERL™ Blood Bank Saline, buffered to a pH of 7.0 -7.2 to match the pH of human blood. Saline is available in Cubitainer™ containers with spigots for ease of use.

  • Phosphate buffered
  • Preservative-free
  • For in vitro diagnostics and laboratory use only, in immunohematological procedure or as a general laboratory diluent when “normal” or isotonic saline is indicated
  • Not for infusion, injection or irrigation

Specifications

Certifications/Compliance Certificates of Analysis are available online for this product
Content And Storage 15°C to 30°C
Type Saline, Buffered
For Use With (Application) Phosphate-buffered saline intended for use in immunohematological procedure or as a general laboratory diluent when normal or isotonic saline is indicated.
Packaging Type Cubitainer
Quantity 1 x 10 L
Stability 12 Months

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.