missing translation for 'onlineSavingsMsg'
Learn More

Neratinib

Catalog No. AC464702500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464702500 250 mg
AC464700010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464702500 Supplier Thermo Scientific Chemicals Supplier No. 464702500
Only null left

CAS: 698387-09-6 | C30H29ClN6O3 | 557.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 698387-09-6
Molecular Formula C30H29ClN6O3
Molecular Weight (g/mol) 557.05
InChI Key JWNPDZNEKVCWMY-UHFFFAOYSA-N
IUPAC Name N-[4-({3-chloro-4-[(pyridin-2-yl)methoxy]phenyl}amino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
SMILES CCOC1=C(NC(=O)C=CCN(C)C)C=C2C(NC3=CC(Cl)=C(OCC4=CC=CC=N4)C=C3)=C(C=NC2=C1)C#N

Specifications

Quantity 250 mg
Formula Weight 557.05
Chemical Name or Material Neratinib
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Suspected of damaging the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only