missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine 98.0+%, TCI America™
$249.78 - $846.29
Chemical Identifiers
| CAS | 2389-60-8 |
|---|---|
| Molecular Formula | C19H28N2O6 |
| Molecular Weight (g/mol) | 380.44 |
| MDL Number | MFCD00062271 |
| InChI Key | DYSBKEOCHROEGX-GGYSOQFKNA-N |
| Synonym | z-lys boc-oh, n-cbz-n'-boc-l-lysine, nepsilon-boc-nalpha-cbz-l-lysine, cbz-lys boc-oh, z-lysine boc-oh, z-l-lys boc-oh, s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-cbz-n-epsilon-boc-l-lysine, in2-benzyloxycarbonyl-n6-tert-butoxycarbonyl-l-lysine, 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid |
| PubChem CID | 98765 |
| IUPAC Name | (2S)-2-{[(benzyloxy)carbonyl]amino}-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 2389-60-8 | |
| 380.44 | |
| DYSBKEOCHROEGX-GGYSOQFKNA-N | |
| 98765 | |
| CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O |
| C19H28N2O6 | |
| MFCD00062271 | |
| z-lys boc-oh, n-cbz-n'-boc-l-lysine, nepsilon-boc-nalpha-cbz-l-lysine, cbz-lys boc-oh, z-lysine boc-oh, z-l-lys boc-oh, s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-cbz-n-epsilon-boc-l-lysine, in2-benzyloxycarbonyl-n6-tert-butoxycarbonyl-l-lysine, 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
Specifications
| 2389-60-8 | |
| White | |
| MFCD00062271 | |
| z-lys boc-oh, n-cbz-n'-boc-l-lysine, nepsilon-boc-nalpha-cbz-l-lysine, cbz-lys boc-oh, z-lysine boc-oh, z-l-lys boc-oh, s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n-cbz-n-epsilon-boc-l-lysine, in2-benzyloxycarbonyl-n6-tert-butoxycarbonyl-l-lysine, 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid | |
| CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 380.44 | |
| 380.44 | |
| Crystalline Powder |
| 62°C | |
| C19H28N2O6 | |
| 5 g | |
| DYSBKEOCHROEGX-GGYSOQFKNA-N | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-6-{[(tert-butoxy)carbonyl]amino}hexanoic acid | |
| 98765 | |
| ≥98.0% (HPLC,T) | |
| Nepsilon-(tert-Butoxycarbonyl)-Nalpha-carbobenzoxy-L-lysine |
Safety and Handling
TSCA : Yes