missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Formyl-L-lysine 98.0+%, TCI America™
Supplier: TCI America F01361G
Specifications
| Nepsilon-Formyl-L-lysine | |
| White | |
| MFCD00037362 | |
| KLPJXDPPMSJWKI-LURJTMIESA-N | |
| (2S)-2-amino-6-formamidohexanoic acid | |
| 174.2 | |
| CHEBI:21884 | |
| ≥98.0% (T) |
| 1190-48-3 | |
| C7H14N2O3 | |
| H-Lys(For)-OH | |
| C(CCNC=O)CC(C(=O)O)N | |
| 1 g | |
| 70923 | |
| 174.20 | |
| Crystalline Powder |
Chemical Identifiers
| 1190-48-3 | |
| 174.2 | |
| KLPJXDPPMSJWKI-LURJTMIESA-N | |
| 70923 | |
| (2S)-2-amino-6-formamidohexanoic acid |
| C7H14N2O3 | |
| MFCD00037362 | |
| H-Lys(For)-OH | |
| CHEBI:21884 | |
| C(CCNC=O)CC(C(=O)O)N |
Safety and Handling
TSCA : No