missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Carbobenzoxy-Nalpha-tosyl-L-lysine 98.0+%, TCI America™

Catalog No. p-7124583
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07351G 1 g
C07355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07351G Supplier TCI America Supplier No. C07351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34235-82-0
Molecular Formula C21H26N2O6S
Molecular Weight (g/mol) 434.507
MDL Number MFCD00059201
InChI Key XSWGRGGVKUEBSE-IBGZPJMESA-N
Synonym Nepsilon-Cbz-Nalpha-tosyl-L-lysine, Tos-Lys(Z)-OH
PubChem CID 14082453
IUPAC Name (2S)-2-[(4-methylphenyl)sulfonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCCCNC(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 434.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-Carbobenzoxy-Nalpha-tosyl-L-lysine